Merck
Z364509
Sigma-Aldrich
Nalgene® media bottles, style 2019
capacity 250 mL
Synonym(e):
bottle nalgene, nalgene containers, sterile bottles
Größe auswählen
48 EA
CHF 383.00
CHF 383.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
48 EA
CHF 383.00
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.17
CHF 383.00
Check Cart for Availability
Materialien
high-density polyethylene cap
square bottle
transparent PETG
Verbindung
Joints threaded neck (38-430mm)
Beschreibung
Not autoclavable
Sterilität
sterile; γ-irradiated
Leistungsmerkmale
graduations
with cap
Verpackung
pack of 48 ea (2 x pack of 24)
Hersteller/Markenname
Nalgene® 2019-0250
Kapazität
250 mL
Allgemeine Beschreibung
  • Nalgene® media bottles are made from rigid and sturdy polyethylene terephthalate copolyester, glycol-modified (PETG).
  • Designed for storing buffers, cultures, and culture media for various laboratory applications.
  • Features a leakproof closure for sample protection.
  • Heat-shrink band around closure and neck facilitate tamper-evident seal.
  • Ideal for refrigerated/frozen storage down to -40°C.
Media bottles (square), with white HDPE screw closure. Low gas permeability, non-pyrogenic. Heat-shrink band around closure and neck provides tamper-evident seal.
Leistungsmerkmale und Vorteile
  • Avoids depyrogenation, expensive washing, and autoclaving steps
  • Sterile and non-pyrogenic
  • Obtained in low particulate format
  • Validated by extensive documentation
  • Useful for storing and sampling active pharmaceutical ingredients and bulk intermediates
  • Suitable for preparation and containment of buffers, cultures or extended storage of pH-sensitive liquids such as culture media.
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)