Merck
Z363545
Sigma-Aldrich
Rührstäbchen, Küvette für Spektroskopie
provides rapid horizontal and vertical mixing, pkg of 1 ea
Synonym(e):
magnetic bar, magnetic bars, stir bars
Größe auswählen
1 EA
CHF 26.40
CHF 26.40
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 26.40
About This Item
UNSPSC-Code:
41122401
NACRES:
SB.32
CHF 26.40
Check Cart for Availability
Verpackung
pkg of 1 ea
Hersteller/Markenname
Bel-Art F37150-0000
Allgemeine Beschreibung
Sized to fit standard 10 mm cuvettes. Provides rapid horizontal and vertical mixing with a minimum of vortex formation and aeration. PTFE coating will not scratch cuvette, 9 mm diameter × 8 mm high.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Andrew M Fales et al.
Biomedical optics express, 10(4), 1919-1934 (2019-05-08)
Innovative biophotonic modalities such as photoacoustic imaging (PAI) have the potential to provide enhanced sensitivity and molecule-specific detection when used with nanoparticles. However, high peak irradiance levels generated by pulsed lasers can lead to modification of plasmonic nanoparticles. Thus, there
Andrew M Fales et al.
Scientific reports, 7(1), 15704-15704 (2017-11-18)
The rapid growth of gold nanoparticle applications in laser therapeutics and diagnostics has brought about the need for establishing innovative standardized test methods for evaluation of safety and performance of these technologies and related medical products. Furthermore, given the incomplete
Questions
  1. diameter of cuvette stirring bar Z363545
    1 answer
    1. The diameter of the cuvette stirring bar is 9 x 8mm (D x H).