Z350400
Carestream® BioMax® MR-Film
Synonym(e):
autoradiography film, xray film
Alle Fotos(1)
About This Item
UNSPSC-Code:
41105341
NACRES:
NB.22
Empfohlene Produkte
Größe
8 in. (20 cm) × 10 in. (25 cm)
Allgemeine Beschreibung
Einschichtfilm für maximale Auflösung und den hellsten Hintergrund. Der empfindlichste Film für direkte 14C-, 35S- und 33P-Autoradiographie. Höhere Auflösung als mit X-OMAT AR und BioMax MS bei direkter und Folienbelichtung sowie Chemilumineszenz für 32P, jedoch mit geringerer Empfindlichkeit.
Each sheet of film is sealed in a light-tight envelope
Rechtliche Hinweise
BioMax is a registered trademark of Carestream Health, Inc.
Carestream is a registered trademark of Carestream Health, Inc.
Kodak is a registered trademark of Eastman Kodak Company
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Shujuan Gao et al.
Journal of molecular signaling, 6, 8-8 (2011-08-13)
The family of A-kinase-anchoring proteins, AKAPs, constitutes a group of molecular scaffolds that act to catalyze dynamic interactions of protein kinase A, protein kinase C, tyrosine kinases, G-protein-coupled receptors and ion channels. AKAP5 (MW ~47 kDa) and AKAP12 (MW ~191
Qian Kong et al.
Experimental and therapeutic medicine, 17(5), 3598-3606 (2019-04-17)
Acute lung injury (ALI) is a critical syndrome that is associated with high morbidity and mortality rates. The activation of the Fas/Fas ligand (FasL) signaling pathway may be an important pathophysiological mechanism during ALI development. Penehyclidine hydrochloride (PHC) has been
Atsuko Kimura et al.
The European journal of neuroscience, 30(2), 299-306 (2009-07-21)
The 5-HT(2C) receptor has been implicated in mood and eating disorders. In general, it is accepted that 5-HT(2C) receptor agonists increase anxiety behaviours and induce hypophagia. However, pharmacological analysis of the roles of these receptors is hampered by the lack