Merck
TMO312089-0004
Nalgene® narrow-mouth bottle
natural high-density polyethylene, size 125 mL, case of 500 ea
Synonym(e):
Nalgene bottle, plastic bottle
Größe auswählen
500 EACH
CHF 454.00
CHF 454.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
500 EACH
CHF 454.00
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
CHF 454.00
Check Cart for Availability
Materialien
natural high-density polyethylene
Verpackung
case of 500 ea
Hersteller/Markenname
Nalgene 312089-0004
Größe
125 mL
Allgemeine Beschreibung
Store and ship light-sensitive reagents with Thermo Scientific Nalgene Narrow-Mouth Opaque Amber HDPE Packaging Bottles with Closure. Ideal for diagnostics, biologicals, specialty chemicals, reagents, adhesives and veterinary pharmaceuticals, these high density polyethylene bottles are injection blow-molded for uniform wall thickness and strength. They meet USP light transmission standards and are bulk packed for packaging applications.
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)