Merck
S4897
Sigma-Aldrich
Sharps safety pouch stand
supports pouch with mouth open for easy access, pkg of 1 ea
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.35
Verpackung
pkg of 1 ea
Hersteller/Markenname
Bel-Art F13236-0000
Allgemeine Beschreibung
Welded steel wire stand, epoxy coated, supports pouch with mouth open for easy access.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)