Merck
IST110078LR
Gas PermASeal heat sealable seal
LabRoll, W × L 78 mm × 200 mm, pack of 1 roll
Synonym(e):
Gas PermASeal heat sealable seal, gas permeable seal, plate sealing film, sample incubation seal
About This Item
UNSPSC-Code:
41122410
NACRES:
NB.15
Sterilität
non-sterile
Verpackung
pack of 1 roll
Hersteller/Markenname
IST Scientific IST-110-078LR
Methode(n)
PCR: suitable
B × L
78 mm × 200 mm
Allgemeine Beschreibung
Non-woven gas permeable peelable seal for cell & seed culture.
Features: Opaque, non-woven, porous, gas permeable, barrier to solid contaminants, pierceable, peelable. Seals to polypropylene and polystyrene plates. Pierceable. Peelable. Not certified free from nucleases and DNA.
Compatibility: Polypropylene (PP), Polystyrene (PS).
Seal integrity range: -20°C to 80°C.
Anwendung
Short term incubation,agriculture & seed storage, cell culture, for use in autoclaves
Rechtliche Hinweise
Gas PermASeal is a trademark of IST Scientific
LabRoll is a trademark of IST Scientific