Merck
HS120648
Reagent Reservoir
capacity 100 mL, polystyrene, sterile; electron beam irradiated, pack of 100 (individually wrapped)
Synonym(e):
Reagent Container, Reagent Storage
Größe auswählen
100 EACH
CHF 177.00
CHF 177.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
100 EACH
CHF 177.00
About This Item
UNSPSC-Code:
41121522
NACRES:
NB.01
CHF 177.00
Check Cart for Availability
Materialien
V-bottom bottom
polystyrene
Sterilität
sterile; electron beam irradiated
Verpackung
pack of 100 (individually wrapped)
Hersteller/Markenname
Heathrow Scientific 120648
Kapazität
100 mL
Allgemeine Beschreibung
Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures. Graduation markingson the top edge and inner walls allows measurements of liquid to be easily identified and reduces waste of expensive reagents. Pour-off spouts design on all four corners enables easy spill-free pouring and reduces chance of dripping.
Leistungsmerkmale und Vorteile
  • Trough design with a “v” shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
  • Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
  • Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
  • Designed to stay flat on the benchtop and not warp
  • Sealed in easy-to-open bags. No scissors needed to open
  • Disposable
  • Design Patent Pending
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Questions
  1. Is this product working with multi-channel pipett? I cant find the dimentions. BR/Josefin
    1 answer
    1. This product is manufactured by Heathrow Scientific, part number 120648 (found in the "Properties" section of the product page). According to the manufacture, this product is designed for multi-channel pipettes. The interior dimensions of this product are as follows: 4.3 x 1.9 x 1.3 in (10.9 x 4.8 x 3.2 cm). More information can be found at this link:
      https://www.heathrowscientific.com/reagent-reservoirs