Merck
GN781090
Greiner CELLSTAR® microplate
384 well, TC treated, black μClear®
Synonym(e):
384 well microplate, 384 well plate, cell culture plate, treated microplate
About This Item
UNSPSC-Code:
41122107
Materialien
black
Verpackung
pack of 120 ea
Hersteller/Markenname
Greiner 781090
Verfügbarkeit
available only in China
Größe
384 wells
Eignung
suitable for (cell culture)
Allgemeine Beschreibung
  • Greiner CELLSTAR® 384 Well Cell Culture Plates enable improved cell adhesion through physical surface treatment.
  • Features a lid that facilitates gas exchange with the lowest possible evaporation and significantly reduces the risk of cross-contamination.
  • Free of detectable DNase, RNase, human DNA, non-pyrogenic and non-cytotoxic.
  • μClear® bottom is a transparent thin-film bottom ideal for cell culture and microscopic applications using fluorescence or luminescence detection methods.
  • A rounded square well profile facilitates flexible working volume and prevents bubbling.
  • Suitable for all common instruments and compatible with automated systems.
  • Alphanumeric well coding.
Rechtliche Hinweise
μClear is a registered trademark of Greiner Bio-One GmbH
CELLSTAR is a registered trademark of Greiner Bio-One GmbH
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)