Merck
EPF740520035
Eppendorf® CryoCube® F740hi
cooling, 5 touchscreen interface, AC/DC input 120 - 208 V, 60 Hz, capacity 740 L
Synonym(e):
CryoCube® F740hi, CryoCube® F740hi-ULT Freezer
About This Item
UNSPSC-Code:
41103034
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Leistungsmerkmale
cooling 5 touchscreen interface
Kapazität
740 L
Allgemeine Beschreibung
The Eppendorf CryoCube F740 series is one of the newest additions to the Eppendorf ultra-low temperature freezer family (also called ultra-low temperature freezer, -80C). The CryoCube F740 series of ultra-low temperature freezers combines the traditionally high quality of our previous ultra-low temperature freezers with a higher capacity to safely store more of your samples while optimizing energy consumption. By incorporating the PhysioCare Concept into the development, the CryoCube F740 is lighter and more comfortable to use, supports ergonomics in the workplace and enables an optimal workflow in the laboratory.
Rechtliche Hinweise
CryoCube is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)