Merck
EP5365000109
Eppendorf® Adapter 25 mL for SmartBlock(TM) 50 mL
Set consisting of 4 adapters and removal tool
Größe auswählen
4 EACH
CHF 191.00
CHF 191.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
4 EACH
CHF 191.00
About This Item
UNSPSC-Code:
41121703
NACRES:
NB.01
CHF 191.00
Check Cart for Availability
Leistungsmerkmale
Set consisting of 4 adapters and removal tool
Verpackung
case of 4 ea
Hersteller/Markenname
Eppendorf® 5365000109
Verwandte Kategorien
Allgemeine Beschreibung
hese individually sensor-controlled SmartBlocks are the best solution for maintaining accurate, uniform temperatures. A wide variety of SmartBlocks for all common lab vessels support a broad range of applications - for use with Eppendorf ThermoMixer C (heating/ cooling and mixing) and ThermoStat C (heating/ cooling).
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)