Merck
CLS480141
Corning® LSE Adapters
for 5/7 mL tubes in 50 mL rotor, 6/cs
Synonym(e):
centrifuge rotor tube adapters, centrifuge tube adapters
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.15
Verpackung
case of 6
pack of 6
Hersteller/Markenname
Corning 480141
Eignung
suitable for (for use with Corning LSE compact centrifuge)
Verwandte Kategorien
Allgemeine Beschreibung
These are optional adapters for Corning LSE compact centrifuge rotors: CLS480136, CLS480137, CLS480138; CLS480139
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
LSE is a trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)