Merck
CLS400084
Support rod for Corning® hot plates and stirrers
stainless steel
About This Item
UNSPSC-Code:
41122800
NACRES:
NB.73
Materialien
stainless steel
Hersteller/Markenname
Corning 400084
Durchm. × L
5/16 in. × 12 in.
Verwandte Kategorien
Allgemeine Beschreibung
These 5/16 x 12 inch stainless steel support rods screw into the base of Corning PC-200/210/220 series hot plate/stirrers.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)