Merck
CLS3081250CO
Pyrex® large knob top desiccators
replacement cover for 10.5L large knob top desiccator, 1/cs
Synonym(e):
desiccator covers, desiccator lids
About This Item
UNSPSC-Code:
41104510
NACRES:
NB.43
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Materialien
borosilicate glass
Verpackung
case of 1
pack of 1
Hersteller/Markenname
Corning 3081-250CO
Eignung
suitable for (general analytical work)
Verwandte Kategorien
Allgemeine Beschreibung
Note: This is the desiccator cover replacement for the Pyrex desiccator CLS3181250.
The Pyrex large knob top desiccators are designed for general analytical work. The large sturdy knob makes the cover easy to handle. Warning: Not for use with vacuum applications. Do not subject to heat or pressure.
Rechtliche Hinweise
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)