Merck
BAF198080000
Bel-Art® Centrifuge Tubes, Rimless
tube capacity × L 90 mL × 162 mm, polypropylene, pack of 12 tubes
Synonym(e):
round bottom tube
About This Item
UNSPSC-Code:
41121703
NACRES:
NB.22
Materialien
polypropylene
round bottom
Leistungsmerkmale
autoclavable
Verpackung
pack of 12 tubes
Hersteller/Markenname
Bel-Art F19808-0000
Röhrchenkapazität × L
90 mL × 162 mm
Verwandte Kategorien
Rechtliche Hinweise
Bel-Art is a registered trademark of SP Scienceware, a division of SP