Merck
Z756997
Kern DBS moisture analyzer, for samples up to 60 g
AC/DC input 110 V AC
Synonym(e):
moisture sensor, moisture tester
About This Item
UNSPSC-Code:
41111502
Hersteller/Markenname
Kern DBS 60-3 US
Messbereich Wiegekapazität
60 g
L × B × H
202 mm × 336 mm × 157 mm
Allgemeine Beschreibung
  • Halogen quartz glass heater
  • Observation window over the sample
  • Internal memory for automatic sequence of 10 drying programs
  • Sample description for up to 99 samples
  • Date and time display as standard
  • 10 sample plates included
Ähnliches Produkt
Produkt-Nr.
Beschreibung
Preisangaben
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)