Merck
Z743187
Julabo® bath covers
plexiglas cover to fit CORIO CD-1001F
Größe auswählen
1 EA
CHF 500.00
CHF 500.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 500.00
About This Item
UNSPSC-Code:
41103706
NACRES:
NB.77
CHF 500.00
Check Cart for Availability
Verpackung
pack of 1 ea
Hersteller/Markenname
Julabo 9970247
Allgemeine Beschreibung
Transparent Plexiglas hood reduces the evaporation of the bath fluid and saves energy. The rising steam is led back into the bath. This means that the bath fluid remains in the bath and refill cycles are extended. An additional benefit is that the hood prevents contamination of the samples in the bath. The hood can be used in the temperature range of -10°C to +80°C.
Rechtliche Hinweise
Julabo is a registered trademark of Julabo USA, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)