Merck
Z682691
Ace drying flask, pear-shaped, with side indents
capacity 1000 mL, joint: ST/NS 29/32, glass
Synonym(e):
Aldrich® drying flask, pear-shaped, with side indents
Größe auswählen
1 EA
CHF 166.00
CHF 166.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 166.00
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
CHF 166.00
Check Cart for Availability
Materialien
glass
pear-shaped flask
single-neck
Verbindung
joint: ST/NS29/32
Sterilität
non-sterile
Leistungsmerkmale
closure type Standard Taper Joint
Verpackung
case of 1
Hersteller/Markenname
Büchi equivalent, 00453
Kapazität
1000 mL
Allgemeine Beschreibung
These pear shaped drying flasks for rotary evaporators have side indents which increase the mixing action when the flask is rotated on an evaporator .
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)