Merck
Z682128
Ace cold trap outer body only for condenser "C" assembly
glass, fits Büchi® models R-200/R-205 and R-114/R-144
Größe auswählen
1 EA
CHF 1’020.00
CHF 1’020.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 1’020.00
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
CHF 1’020.00
Check Cart for Availability
Materialien
glass
Hersteller/Markenname
Büchi equivalent, 40641
Verwandte Kategorien
Allgemeine Beschreibung
The cold trap outer body for the Ace condenser "C" assembly, is glass with a 35/20 inner spherical joint bottom connection that attaches to the receiver flasks. Features standard top and side flange that connects directly to the Büchi rotary evaporator.
Rechtliche Hinweise
Büchi is a registered trademark of Büchi Labortechnik AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)