Merck
Z678155
Finnpipette® F3 variable volume pipette
volume 1-10 mL
Größe auswählen
1 EA
CHF 384.00
CHF 384.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 384.00
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
CHF 384.00
Check Cart for Availability
Leistungsmerkmale
color-coded red
electronic: no
single-channel
Verpackung
pack of 1 ea
Hersteller/Markenname
Finn 4640070
Volumen
1-10 mL
Allgemeine Beschreibung
Finnpipette F3 has a double action pipetting button with rotating upper part enabling smooth and steady pipetting action at the same time as preventing accidental volume alterations. Finpipette F3 is equipped with the Advanced Volume Gearing (AVG). This is a self-supporting modular volume adjustment mechanism separated from the pipette body. This improves accuracy and precision by preventing possible effects of hand warmth on the accuracy of measurements.
  • Double action pipetting button with very light and smooth pipetting action
  • Advanced Volume Gearing mechanism (AVG) ensures increased accuracy and precision
  • Big display with white numbers on black background for increased visibility
  • Color coded for easy tip volume recognition
Rechtliche Hinweise
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)