Merck
Z557412
JULABO COOLING COIL FOR SHAKING WATER BATH
pack of 1 ea
Größe auswählen
1 EA
CHF 271.00
CHF 271.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 271.00
About This Item
UNSPSC-Code:
41103025
NACRES:
NB.77
CHF 271.00
Check Cart for Availability
Verpackung
pack of 1 ea
Hersteller/Markenname
Julabo 8970416
Allgemeine Beschreibung
Cooling coil for retrofitting shaking water baths SW22/SW23. The cooling coil is screwed into the water bath through the drain tap. Once installed, the bath may be operated near or slightly below the ambient temperature.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)