Merck
Z551228
Graduated Valumetric bottles
black phenolic cap (with rubber liner), capacity 60 mL (2 oz)
Synonym(e):
W216979
Größe auswählen
48 EA
CHF 216.00
CHF 216.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
48 EA
CHF 216.00
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.17
CHF 216.00
Check Cart for Availability
Materialien
black phenolic cap (with rubber liner)
clear bottle
round bottle
soda-lime glass bottle (Type III)
wide-mouth bottle
Verbindung
Joints threaded neck (38-400)
Leistungsmerkmale
with cap
Verpackung
pkg of 48 ea
Hersteller/Markenname
WHEATON® W216979
Kapazität
60 mL (2 oz)
Durchm. × H
42 mm × 83 mm
Allgemeine Beschreibung
Ideal for use as sample bottles. They are also useful for dry powders, soil samples, and specimen storage. All bottles are made of clear, Type III soda lime glass.
Leistungsmerkmale und Vorteile
  • Clear, USP Type III soda-lime glass
  • Graduated in mL and ounces
  • Easy removal of contents
  • Available with caps attached or bulk packs without caps
  • HAZCOM labels, provided in each case of bottles with the exception of bulk packs, allow for ID of content and safety notification
  • Tamper Evident Seal keeps bottle sealed, protecting contents from accidental misuse (not included in bulk packs)
Rechtliche Hinweise
Valumetric is a trademark of Wheaton Industries, Inc.
WHEATON is a registered trademark of Wheaton Industries, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)