Merck
SYNF602015F
Synthware Buchner funnel
15 mL, porosity: fine
Synonym(e):
Synthware F602015F, filtering funnel
Größe auswählen
1 EA
CHF 45.80
CHF 45.80
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 45.80
About This Item
UNSPSC-Code:
41121809
NACRES:
NB.51
CHF 45.80
Check Cart for Availability
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F602015F
Kapazität
15 mL
15 mL
Allgemeine Beschreibung
With sealed-in glass disc of the listed porosity. Not recommended for filtration where pressure would exceed 15psi. XC-extra coarse, C-Coarse, M-Medium, F-Fine porosity.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)