Merck
Z248320
J-KEM® thermocouple probe
Single-element, thermocouple type, T
About This Item
UNSPSC-Code:
41112303
NACRES:
NB.76
Beschreibung
PTFE-coated
Leistungsmerkmale
thermocouple type T
Hersteller/Markenname
J-Kem
Größe
1/4 in. × 12 in.
Rechtliche Hinweise
J-KEM is a registered trademark of J-KEM Electronics, Inc.