Merck
Z153923
Sigma-Aldrich
Deckel für Duran® Laborflaschen
Caps for Duran® laboratory bottles
1 of 1 reviewers received a sample product or took part in a promotion
GL 45 neck thread, blue polypropylene (with lip seal)
Synonym(e):
caps bottles, glass bottles caps
Größe auswählen
10 EA
CHF 24.10
CHF 24.10
Check Cart for Availability
Größe auswählen
Ansicht ändern
10 EA
CHF 24.10
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.17
CHF 24.10
Check Cart for Availability
Materialien
blue polypropylene (with lip seal)
Hersteller/Markenname
DURAN®
Schott
Parameter
-40-140 °C temp. stability
Fitting
GL 45 neck thread
Allgemeine Beschreibung
The classic laboratory bottle GL threaded screw cap features the tried and tested cap plug sealing system ensuring a liquid tight seal. Available in four colours; blue, green, yellow and grey with matching pouring rings. Distinguishing media types is simplified and interchanging of screw caps and the risk of cross contamination is greatly reduced.
Leistungsmerkmale und Vorteile
  • Cap with integral lip seal provides effective sealing for short term storage
  • Pouring ring for drip-free pouring
  • The colour helps prevent interchanging of cap mix-ups and reduces the risk of cross contamination
  • Fully compatible with the GL 45 threads and therefore highly versatile
Rechtliche Hinweise
DURAN is a registered trademark of DWK Life Sciences
Empfehlung
Produkt-Nr.
Beschreibung
Preisangaben
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Artikel
A 4-step comprehensive ICP-MS workflow including sample homogenization, with standard addition calibration method, to determine heavy metals in Cannabis sativa.
Questions
  1. It says caps with lip seal - is this the rings that go around the top of the bottle to make pouring easier? We ordered these caps and they did not come with those
    1 answer
    1. A lip seal is lip or ring that is molded into the top of the cap. It fits against the interior wall of the tube or bottle preventing the contents from coming in contact with the seal or threads, thereby reducing the chances of sample contamination. See the link below for Duran pouring rings which are offered separately. There are several GL45 options:
      https://www.sigmaaldrich.com/substance/pouringringsforduranlaboratorybottles1234598765
      Helpful?
Reviews
1 of 1 reviewers received a sample product or took part in a promotion
Active Filters
  1. La Jolla, CA
    • Reviews 2
    • Votes 0
    5 out of 5 stars.
    Replacement caps. I would buy again.
    Good replacement caps for the ones that are falling to bits.