Merck
Z101214
Sigma-Aldrich
Aldrich® Inline-Blasenzähler
capacity 5-7 mL
Synonym(e):
bubblers, in-line bubblers, scientific bubblers
Größe auswählen
1 EA
CHF 129.00
CHF 129.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 129.00
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
CHF 129.00
Check Cart for Availability
Kapazität
5-7 mL
Verwandte Kategorien
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Questions
  1. Dear Sigma Aldrich team, can you please specify the dimensions of this bubbler? Especially the diameter of the inner and outer tubes.
    1 answer
    1. The height of the bubbler is 205 MM, the T-cross has an outer diameter of 8 MM and a inner diameter of 5.2 MM. The side arm has an outer diameter of 8 MM and a inner diameter of 4.8 MM.