Merck
TEM-3750
Sigma-Aldrich
CryoCapsule
3 mm, pack of 10
About This Item
UNSPSC-Code:
41111761
Verpackung
pack of 10
Allgemeine Beschreibung
The CryoCapsule is a new tool that is compatible with High Pressure Freezing (HPF). The CryoCapsule is dedicated to correlative light to electron microscopy experiments. Comparable to a small petri dish, it is composed of a landmarked sapphire disc and
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)