SYNV121000
Synthware™ volume vacuum trap
1000 mL, joint: ST/NS 45/50
Synonym(e):
Synthware V121000
Empfohlene Produkte
Verbindung
joint: ST/NS45/50
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware V121000
Kapazität
1000 mL
Verwandte Kategorien
Allgemeine Beschreibung
Trap is designed for large volume solvent removal by vacuum. With 45/50 joint. Connections at center tube is 12 mm O.D. while side tube is 10 mm.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.