Merck
SYNS734040
Synthware sublimator, complete
40 mL, complete
Synonym(e):
Synthware S734040, sublimation apparatus
Größe auswählen
1 EA
CHF 351.00
CHF 351.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 351.00
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.51
CHF 351.00
Check Cart for Availability
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware S734040
Kapazität
40 mL
Allgemeine Beschreibung
A grease-free, two-piece sublimation apparatus consisting of an upper water cooled condenser and lower vessel to hold the material to be sublimed. The sublimate collects on the lower end of the condenser which has a flat bottom. Condenser is constructed using a grease-free, threaded valve. Connection of the condenser and lower vessel via an O-ring joint enables the two parts to be gently separated for maximum product recovery. Complete apparatus consists of the cold finger condenser, lower vessel, Viton® O-ring and stainless steel pinch clamp.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)