Merck
SYNM260003
Synthware all-glass vacuum/inert gas manifold with solid high vacuum stopcocks
number of ports, 3, Hose Connections: Front-left-right, Rear-left
Synonym(e):
Synthware M260003, vacuum manifold, vacuum unit
Größe auswählen
1 EA
CHF 645.00
CHF 645.00
Voraussichtliches Versanddatum02. Juni 2025
Größe auswählen
Ansicht ändern
1 EA
CHF 645.00
About This Item
UNSPSC-Code:
40161501
NACRES:
NB.51
CHF 645.00
Voraussichtliches Versanddatum02. Juni 2025
Leistungsmerkmale
number of ports 3
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware M260003
Allgemeine Beschreibung
With 4 mm double oblique, high vacuum solid glass stopcocks precision ground with corresponding numbers on barrel and plug. Solid glass stopcocks have constant pressure retention devices. Beams are 22 mm O.D. with front-left-right and rear-left 10 mm hose connections. Distance between stopcocks is 75 mm center to center.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)