Merck
SYNM350004
Synthware double manifold with left-hand side SJ 35/20 joint
joint: ST/NS 35/20, Ports: 4
Synonym(e):
Synthware M350004, vacuum manifold, vacuum unit
Größe auswählen
1 EA
CHF 661.00
CHF 661.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 661.00
About This Item
UNSPSC-Code:
40161501
NACRES:
NB.51
CHF 661.00
Check Cart for Availability
Verbindung
joint: ST/NS35/20
Verpackung
pack of 1 ea
Hersteller/Markenname
M350004
Allgemeine Beschreibung
Synthware glass manifold featuring high vacuum hollow glass stopcocks, the smaller O.D. 17 mm front tube is designed for inert gases, while the larger O.D. 22 mm back tube is for vacuum, which has a 35/20 socket on the left-hand side. Hose connections are located front-left-right.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)