SYNM202404
Synthware™ vacuum manifold with high vacuum valves
number of ports, 4, Hose Connection: right
Synonym(e):
Synthware M202404, vacuum manifold, vacuum unit
Empfohlene Produkte
Leistungsmerkmale
number of ports 4
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware M202404
Allgemeine Beschreibung
With a serrated hose connection(10 mm O.D.) at right hand side. Main tube is 22 mm O.D. and the distance between valves is 100 mm center to center.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.