Merck
SYNF419100
Synthware two neck round bottom flask with angled side neck
100 mL, center joint: ST/NS 24/40, side joint: ST/NS 19/22
Synonym(e):
Synthware F419100, flask, two-neck flask, round bottom flask
Größe auswählen
1 EA
CHF 70.70
CHF 70.70
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 70.70
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.51
CHF 70.70
Check Cart for Availability
Materialien
borosilicate glass flask
round bottom
two-neck (angled side neck)
Verbindung
center joint: ST/NS24/40
side joint: ST/NS19/22
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F419100
Kapazität
100 mL
100 mL
Allgemeine Beschreibung
Round bottom with joints. Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)