Merck
SYNC564225
Synthware coil-style reflux condenser with removable hose connections
Synthware™ coil-style reflux condenser with removable hose connections
joint: ST/NS 24/40, hose size 10 mm
Synonym(e):
Synthware C564225, coil-type condenser
Größe auswählen
1 EA
CHF 231.00
CHF 231.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 231.00
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.51
CHF 231.00
Check Cart for Availability
Verbindung
joint: ST/NS24/40
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware C564225
Schlauchgröße
10 mm
Verwandte Kategorien
Allgemeine Beschreibung
Coil style reflux condenser with tightly wrapped coil providing maximum cooling. Excellent for use with volatile solvents. Condenser has a top outer standard taper joint and lower inner drip tip joint.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Active Filters
  1. 1 Ratings-Only Review