Merck
SYNA371922
Synthware Claisen adapter
joint: ST/NS 19/22
Synonym(e):
Synthware A371922, distillation adapter, glass Claisen adapter
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.51
Verbindung
joint: ST/NS19/22
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A371922
Verwandte Kategorien
Allgemeine Beschreibung
Provides dual entry into a variety of flasks.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.