Merck
S70611
OCTADECANOIC ACID 1-MORPHOLIN-4-YLMETHYL-2-(2-OXO-PYRROLIDIN-1-YL)-ET ESTER, HCL
AldrichCPR
Größe auswählen
25MG
CHF 83.90
CHF 83.90
Voraussichtliches Versanddatum28. Mai 2025
Größe auswählen
Ansicht ändern
25MG
CHF 83.90
About This Item
Lineare Formel:
C29H55ClN2O4
CAS-Nummer:
Molekulargewicht:
531.226
MDL-Nummer:
CHF 83.90
Voraussichtliches Versanddatum28. Mai 2025
Sonstige Hinweise
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)