Merck
HAM62576-01
Foot switch for Hamilton® Microlab® instruments
pack of 1 ea
Größe auswählen
1 EA
CHF 254.00
CHF 254.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 254.00
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
CHF 254.00
Check Cart for Availability
Verpackung
pack of 1 ea
Kabel L
1800 mm
Allgemeine Beschreibung
The Foot Switch is designed to work with Hamilton Microlab instruments. It is for applications where the user′s hands need to be free such as during an animal dosing experiment. The foot switch connects into the probe receptacle and sits on the ground. A simple press of the foot switch communicates with the pump and advances the programmed method.
Rechtliche Hinweise
Hamilton is a registered trademark of Hamilton Co.
Microlab is a registered trademark of Hamilton Co.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)