Merck
HAM60676-01
Universal dispensing valve, for Hamilton® Microlab® 600 pipettor
left valve
Größe auswählen
1 EA
CHF 380.00
CHF 380.00
Voraussichtliches Versanddatum30. Mai 2025
Größe auswählen
Ansicht ändern
1 EA
CHF 380.00
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
CHF 380.00
Voraussichtliches Versanddatum30. Mai 2025
Materialien
PTFE/CTFE
aluminum housing
Leistungsmerkmale
left valve
Parameter
0-100 psig pressure
10-100 °C temperature (50-212 °F)
Fitting
1/4-28 thread
Eignung
suitable for (fits all Microlab 600 instruments)
Allgemeine Beschreibung
The Microlab 600 uses a universal valve that can support single dispensing, dual dispensing, diluting, and continuous dispensing. This valve functions on all Microlab 600 units. The valve has an aluminum housing with a PTFE-rotary plug. Syringes connect into the 5/16-32 threaded port.
Rechtliche Hinweise
Hamilton is a registered trademark of Hamilton Co.
Microlab is a registered trademark of Hamilton Co.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)