Merck
HAM59000-10
Hamilton® Microlab® 600 instrument syringe
model, 1702 BFP, volume 25 μL
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
Materialien
Gastight Borosilicate glass/PTFE syringe
Leistungsmerkmale
model 1702 BFP
Hersteller/Markenname
Hamilton 59000-10
Parameter
10-115 °C temp. range (50-239 °F)
1000 psig pressure
ID × AD
0.729 mm × 7.75 mm
Volumen
25 μL
Rechtliche Hinweise
Hamilton is a registered trademark of Hamilton Co.
Microlab is a registered trademark of Hamilton Co.