Merck
DWK74504-20
KIMBLE® scintillation vials with attached pulp backed foil lined PP cap
lips on vial transparent borosilicate glass bottle, vial capacity (20 mL), screw cap
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.17
Materialien
lips on vial transparent borosilicate glass bottle
metal foil liner
white polypropylene cap
Sterilität
non-sterile
Leistungsmerkmale
class A
with cap
Verpackung
case of 500 ea 5x shrink-wrapped trays of 100 vials
Hersteller/Markenname
Kimble® 74504-20
Flaschendurchm. × H
(27 mm) (57 mm)
Fläschchenkapazität
(20 mL)
Fitting
22-400 neck
22-400
Allgemeine Beschreibung
KIMBLE® glass scintillation vials feature 20mL borosilicate glass vials with polypropylene caps that are securely attached. These vials are packaged in cellular trays that feature alphanumeric coordinates, enabling quick and effortless identification.
Leistungsmerkmale und Vorteile
  • Converted from Type I, 33 expansion borosilicate glass tubing.
  • Each tray contains 100 vials
  • Five trays per case, shrink-wrapped to preserve cleanliness
  • Caps attached
Rechtliche Hinweise
KIMBLE is a registered trademark of DWK Life Sciences
Ähnliches Produkt
Produkt-Nr.
Beschreibung
Preisangaben
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)