BR87818
BRAND® PP beaker with spout, low form
volume 600 mL
Synonym(e):
Beakers, Plastics
Alle Fotos(2)
About This Item
UNSPSC-Code:
41121803
NACRES:
NB.43
Empfohlene Produkte
Materialien
polypropylene
Beschreibung
in addition to ISO 7056
Leistungsmerkmale
graduations
beaded rim and spout
Hersteller/Markenname
BRAND 87818
Volumen
600 mL
Allgemeine Beschreibung
PP, high clarity. ISO 7056. With graduation, beaded rim and spout. Cleaning up to max. 60 °C is recommended to preserve marks and inscriptions.
Sonstige Hinweise
Browse all plastic beakers
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)