Merck
BR87602
BRAND® PP beaker with spout, low form
volume 10 mL
Synonym(e):
Beakers, Plastics
Größe auswählen
12 EA
CHF 22.20
CHF 22.20
Voraussichtliches Versanddatum17. April 2025
Größe auswählen
Ansicht ändern
12 EA
CHF 22.20
About This Item
UNSPSC-Code:
41121803
NACRES:
NB.43
CHF 22.20
Voraussichtliches Versanddatum17. April 2025
Materialien
polypropylene
Beschreibung
ISO 7056
Leistungsmerkmale
graduations
beaded rim and spout
Hersteller/Markenname
BRAND 87602
Volumen
10 mL
Allgemeine Beschreibung
The BRAND® PP Beaker with Spout in low form is crafted from high-clarity polypropylene (PP) and complies with ISO 7056 standards, ensuring durability and reliability for laboratory applications. This beaker features clear graduation markings for precise measurement, along with a beaded rim that enhances strength and provides a comfortable grip. The integrated spout allows for easy pouring, minimizing spills and improving accuracy during liquid transfer. To preserve the marks and inscriptions, it is recommended to clean the beaker at temperatures up to a maximum of 60 °C.
Sonstige Hinweise
Browse all plastic beakers
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)