Merck
BR74511
BRAND® filter funnel support with base plate
for 4 funnels
Synonym(e):
Accessories, Filter funnels, Funnels, Supports
Größe auswählen
1 EA
CHF 207.00
CHF 207.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 207.00
About This Item
UNSPSC-Code:
41121809
NACRES:
NB.24
CHF 207.00
Check Cart for Availability
Materialien
polypropylene
Leistungsmerkmale
for 4 funnels
Hersteller/Markenname
BRAND 74511
L × B
450 mm × 140 mm
Allgemeine Beschreibung
These PP plates feature a stainless steel support rod with dimensions of 12 mm in diameter and 595 mm in length. They are designed to support 2 or 4 funnels with a top outer diameter ranging from 50 mm to 120 mm. The height of the funnels is adjustable, allowing for customized setups based on your specific needs.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)