Merck
BR703459
BRAND® reagent reservoir with lid for multichannel pipettes
capacity 60 mL
Synonym(e):
Accessories, Air displacement pipettes, Air interface pipettes, Microliter pipettes, Multichannel Pipettes, Pipettes, Piston-operated pipettes, Single channel Pipettes
Größe auswählen
10 EA
CHF 24.10
CHF 24.10
Check Cart for Availability
Größe auswählen
Ansicht ändern
10 EA
CHF 24.10
About This Item
UNSPSC-Code:
41121522
NACRES:
NB.01
CHF 24.10
Check Cart for Availability
Materialien
V-bottom bottom
colorless
polypropylene
Sterilität
non-sterile
Verpackung
pack of 10 ea
Hersteller/Markenname
BRAND 703459
Kapazität
60 mL
Allgemeine Beschreibung
The BRAND® Reagent Reservoir with Lid is designed for use with multichannel pipettes, providing a reliable solution for efficient liquid handling in laboratory applications. This reservoir is completely autoclavable at 121 °C (2 bar) in accordance with DIN EN 285 standards, ensuring sterility and safety for sensitive experiments.CERTIFIED LIFE SCIENCE QUALITY: Virgin raw materials, Manufactured under controlled room conditions, Intensive in-process controls, Visual inspections, Batch management, Final product inspection, Proven functionality
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)