Merck
BR36975
BRAND® volumetric flask, USP, BLAUBRAND®
capacity 20 mL, joint: ST/NS 10/19, glass stopper
Synonym(e):
BRAND® volumetric flask BLAUBRAND® USP conformity certified, BLAUBRAND®, USP, Volumetric flasks
Größe auswählen
2 EA
CHF 49.70
CHF 49.70
Check Cart for Availability
Größe auswählen
Ansicht ändern
2 EA
CHF 49.70
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
CHF 49.70
Check Cart for Availability
Materialien
glass flask
glass stopper
Verbindung
joint: ST/NS10/19
Leistungsmerkmale
accuracy class A
Verpackung
pack of 2 ea
Hersteller/Markenname
BRAND 36975
Kapazität
20 mL
Allgemeine Beschreibung
The BRAND® volumetric flask, USP, BLAUBRAND® is constructed from high-quality Boro 3.3 glass and adheres to DIN EN ISO 1042 standards. It is calibrated to contain (TC, In), ensuring precise liquid measurement for various laboratory applications. Each flask is accompanied by a USP batch certificate, providing assurance of compliance with stringent quality standards. This volumetric flask is an essential tool for accurate analytical work, making it suitable for laboratories that require reliable and certified liquid handling solutions.
Rechtliche Hinweise
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)