Merck
BR145505
BRAND® funnel short stem, glass
short stem, borosilicate glass, diam. 35 mm
Synonym(e):
Funnels
Größe auswählen
1 EA
CHF 9.80
CHF 9.80
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 9.80
About This Item
UNSPSC-Code:
41121809
NACRES:
NB.43
CHF 9.80
Check Cart for Availability
Materialien
borosilicate glass
Leistungsmerkmale
for rapid filtration
Hersteller/Markenname
BRAND 145505
Durchmesser
35 mm
Schaftdurchm. × L
6 mm × 35 mm
Verwandte Kategorien
Allgemeine Beschreibung
The BRAND® Funnel with Short Stem is designed for efficient and rapid filtration in laboratory applications. Made from high-quality Boro 3.3 glass, this funnel combines durability with excellent performance. The short stem facilitates quick liquid transfer and minimizes the risk of clogging during filtration processes. The funnel is angled at 60°, optimizing flow rates. The upper rim is either ground or fire-polished, ensuring a smooth finish that enhances safety and ease of use.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)