Merck
BR122574
BRAND® laboratory bottle, graduated with screw cap
capacity 10000 mL
Synonym(e):
Bottles, Glass, Laboratory bottle
Größe auswählen
1 EA
CHF 334.00
CHF 334.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 334.00
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
CHF 334.00
Check Cart for Availability
Materialien
borosilicate glass bottle
clear bottle
narrow-mouth bottle
polypropylene cap
round bottle
Verbindung
Joints threaded neck (GL 45)
Sterilität
non-sterile
Leistungsmerkmale
autoclavable
graduations
with cap
Verpackung
pack of 1 ea
Hersteller/Markenname
BRAND 122574
Kapazität
10000 mL
Durchm. × H
234 mm × 412 mm
Allgemeine Beschreibung
Threaded to DIN ISO 4796. Studded base for stability and abrasion resistance. Graduated, with marking area. Screw cap and pouring ring PP. Autoclavable (121 °C).
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)