Merck
BAF371250078
Spinfin® magnetic stirring bar
H × diam. 0.5 in. (12.7 mm) × 0.875 in. (22.2 mm)
Synonym(e):
Spinfin® magnetic stirring bar
About This Item
UNSPSC-Code:
41122800
NACRES:
NB.51
Materialien
PTFE-coated
Verpackung
pack of 1 ea
Hersteller/Markenname
Bel-Art F37125-0078
H × Durchm.
0.5 in. (12.7 mm) × 0.875 in. (22.2 mm)
Allgemeine Beschreibung
Mit dünnen Flossen oben
Spinfin® magnetic stirring bars agitate liquids in round bottom vessels such as test tubes, cylinders and flasks, where other stirring bars cannot be used.
Leistungsmerkmale und Vorteile
  • Maintains its position in a vessel and will not wobble
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE coating – Superior Chemical Compatibility
  • Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
  • Autoclave safe
Rechtliche Hinweise
Spinfin is a registered trademark of SP Scienceware, a division of SP
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)