Merck
63730
Sigma-Aldrich
2-Mercaptobenzothiazol
technical, ≥90% (T)
Synonym(e):
2-Benzothiazolthiol, MBT
About This Item
Empirische Formel (Hill-System):
C7H5NS2
CAS-Nummer:
Molekulargewicht:
167.25
Beilstein:
119484
EG-Nummer:
MDL-Nummer:
UNSPSC-Code:
12352100
PubChem Substanz-ID:
NACRES:
NA.22
Qualität
technical
Assay
≥90% (T)
Form
solid
mp (Schmelzpunkt)
177-181 °C (lit.)
SMILES String
Sc1nc2ccccc2s1
InChI
1S/C7H5NS2/c9-7-8-5-3-1-2-4-6(5)10-7/h1-4H,(H,8,9)
InChIKey
YXIWHUQXZSMYRE-UHFFFAOYSA-N
Piktogramme
Exclamation markEnvironment
Signalwort
Warning
H-Sätze
Gefahreneinstufungen
Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1
Lagerklassenschlüssel
11 - Combustible Solids
WGK
WGK 2
Flammpunkt (°F)
392.0 °F - closed cup
Flammpunkt (°C)
200 °C - closed cup
Persönliche Schutzausrüstung
dust mask type N95 (US), Eyeshields, Faceshields, Gloves
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Michael Mah et al.
European journal of orthodontics, 36(6), 657-664 (2013-06-19)
To investigate the different effects of changes in the occlusal plane, incisors inclination, and maxillary intercanine width on the curvature of the smiling line. Records of 46 subjects (28 females and 18 males, mean age 16.6 ± 4.2 years) with
H Parham et al.
Journal of hazardous materials, 205-206, 94-100 (2012-01-17)
Mercury in the lowest levels of concentrations is dangerous for human health due to its bioaccumulation in body and toxicity. This investigation shows the effective removal of mercury (II) ions from contaminated surface waters by modified magnetic iron oxide nanoparticles
Chul Jae Lee et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 68(5), 1313-1319 (2007-03-31)
In this study, the adsorption orientation for 2-mercaptobenzothiazole (MBT) and 2-mercaptobenzoxazole (MBO) on to silver mirror and silver sol substrates have been studied by surface enhanced Raman scattering (SERS). The MBT and MBO were chemisorbed on both silver mirror and
Fei Wang et al.
Organic letters, 13(12), 3202-3205 (2011-05-20)
An efficient strategy for the synthesis of a variety of 2-mercaptobenzothiazole derivatives has been developed. The reaction proceeded from o-haloaniline derivatives and carbon disulfide via a tandem reaction in the presence of 1,8-diazabicyclo[5.4.0]undec-7-ene (DBU) to afford the corresponding 2-mercaptobenzothiazole derivatives
H Parham et al.
Journal of hazardous materials, 151(2-3), 636-641 (2007-07-27)
A rapid sensitive and versatile method for simultaneous determination of 2-mercaptobenzothiazole (MBT) and 2-mercaptobezoxazole (MBO) based on the square wave voltammetric (SWV) using mercury drop electrode (SMDE) has been presented. A three-electrode system containing SMDE working electrode, Pt auxiliary electrode