Merck
A Replication-Competent Vesicular Stomatitis Virus for Studies of SARS-CoV-2 Spike-Mediated Cell Entry and Its Inhibition.
Cell host & microbe (2020-08-02)
M Eugenia Dieterle, Denise Haslwanter, Robert H Bortz, Ariel S Wirchnianski, Gorka Lasso, Olivia Vergnolle, Shawn A Abbasi, J Maximilian Fels, Ethan Laudermilch, Catalina Florez, Amanda Mengotto, Duncan Kimmel, Ryan J Malonis, George Georgiev, Jose Quiroz, Jason Barnhill, Liise-Anne Pirofski, Johanna P Daily, John M Dye, Jonathan R Lai, Andrew S Herbert, Kartik Chandran, Rohit K Jangra
RÉSUMÉ
There is an urgent need for vaccines and therapeutics to prevent and treat COVID-19. Rapid SARS-CoV-2 countermeasure development is contingent on the availability of robust, scalable, and readily deployable surrogate viral assays to screen antiviral humoral responses, define correlates of immune protection, and down-select candidate antivirals. Here, we generate a highly infectious recombinant vesicular stomatitis virus (VSV) bearing the SARS-CoV-2 spike glycoprotein S as its sole entry glycoprotein and show that this recombinant virus, rVSV-SARS-CoV-2 S, closely resembles SARS-CoV-2 in its entry-related properties. The neutralizing activities of a large panel of COVID-19 convalescent sera can be assessed in a high-throughput fluorescent reporter assay with rVSV-SARS-CoV-2 S, and neutralization of rVSV-SARS-CoV-2 S and authentic SARS-CoV-2 by spike-specific antibodies in these antisera is highly correlated. Our findings underscore the utility of rVSV-SARS-CoV-2 S for the development of spike-specific therapeutics and for mechanistic studies of viral entry and its inhibition.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
Camostat mesylate, ≥98% (HPLC)
Sigma-Aldrich
Cathepsin L Inhibitor III, The Cathepsin L Inhibitor III controls the biological activity of Cathepsin L. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.