Principaux documents
57479
Raccords Rheodyne®
nut and ferrule for 1/8 in. tubing, pkg of 1 ea
About This Item
Code UNSPSC :
41115700
Matériaux
stainless steel nut and ferrule
Conditionnement
pkg of 1 ea
Raccords
nut and ferrule for 1/8 in. tubing
Produits recommandés
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Informations légales
Rheodyne is a registered trademark of IDEX Health & Science, LLC